EMAIL THIS PAGE TO A FRIEND
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association
A novel autoantibody against moesin in the serum of patients with MPO-ANCA-associated vasculitis.
PMID 24319012
Abstract
Antineutrophil cytoplasmic autoantibody (ANCA) directed against myeloperoxidase (MPO), a diagnostic criterion in MPO-ANCA-associated vasculitis (MPO-AAV), does not always correlate with disease activity. Here, we detected autoantibodies against moesin, which was located on the surface of stimulated endothelial cells, in the serum of patients. The anti-moesin autoantibody titer was evaluated by ELISA. Seventeen kinds of cytokines/chemokines were measured by a Bio-Plex system. Serum creatinine in the anti-moesin autoantibody-positive group was higher than that in the negative group. Additionally, interferon (IFN)-γ, macrophage chemotactic peptide-1 (MCP-1), interleukin (IL)-2, IL-7, IL-12p70, IL-13, granulocyte/macrophage colony-stimulating factor (GM-CSF) and granulocyte colony-stimulating factor were significantly higher in the positive group. Furthermore, IL-7 and IL-12p70 levels correlated with the anti-moesin autoantibody titer. Based on these findings and the binding of anti-moesin IgG to neutrophils and monocytes, we detected the secretion of cytokines/chemokines such as IFN-γ, MCP-1 and GM-CSF from these cells. The anti-moesin autoantibody existed in the serum of patients with MPO-AAV and was associated with the production of inflammatory cytokines/chemokines targeting neutrophils with a cytoplasmic profile, which suggests that the anti-moesin autoantibody has the possibility to be a novel autoantibody developing vasculitis via neutrophil and endothelial cell activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8542 Anti-Human IgG (Fab specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701267 Anti-Human IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701277 Anti-Human IgG (Fc specific), highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A9544 Anti-Human IgG (Fc specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701276 Anti-Human IgG (Fc specific)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
A3312
Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A5403 Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A3187 Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A3150 Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A3188 Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
SAB3701291 Anti-Human IgG (γ-chain specific)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701336
Anti-Human IgG (H+L)-Alkaline Phosphatase antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3701335 Anti-Human IgG (H+L)-Alkaline Phosphatase antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3701334 Anti-Human IgG (H+L)-Alkaline Phosphatase antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
A1543 Anti-Human IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
SAB3701239 Anti-Human IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701250 Anti-Human IgG F(ab′)2, highly cross adsorbed-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701248 Anti-Human IgG F(ab′)2-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701249 Anti-Human IgG F(ab′)2-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
A2064 Anti-Human IgG−Alkaline Phosphatase antibody, Mouse monoclonal, clone GG-5, purified from hybridoma cell culture
SAB3701029 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
A9309 Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701131 Anti-Mouse IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701159 Anti-Mouse IgG (H+L), F(ab) fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701015 Anti-Mouse IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700993 Anti-Mouse IgG F(ab′)2-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder