EMAIL THIS PAGE TO A FRIEND
Pflugers Archiv : European journal of physiology
Endogenous endothelin 1 mediates angiotensin II-induced hypertrophy in electrically paced cardiac myocytes through EGFR transactivation, reactive oxygen species and NHE-1.
PMID 24327206
Abstract
Emerging evidence supports a key role for endothelin-1 (ET-1) and the transactivation of the epidermal growth factor receptor (EGFR) in angiotensin II (Ang II) action. We aim to determine the potential role played by endogenous ET-1, EGFR transactivation and redox-dependent sodium hydrogen exchanger-1 (NHE-1) activation in the hypertrophic response to Ang II of cardiac myocytes. Electrically paced adult cat cardiomyocytes were placed in culture and stimulated with 1xa0nmolxa0l(-1) Ang II or 5xa0nmolxa0l(-1) ET-1. Ang II increased ~45xa0% cell surface area (CSA) and ~37xa0% [(3)H]-phenylalanine incorporation, effects that were blocked not only by losartan (Los) but also by BQ123 (AT1 and ETA receptor antagonists, respectively). Moreover, Ang II significantly increased ET-1 messenger RNA (mRNA) expression. ET-1 similarly increased myocyte CSA and protein synthesis, actions prevented by the reactive oxygen species scavenger MPG or the NHE-1 inhibitor cariporide (carip). ET-1 increased the phosphorylation of the redox-sensitive ERK1/2-p90(RSK) kinases, main activators of the NHE-1. This effect was prevented by MPG and the antagonist of EGFR, AG1478. Ang II, ET-1 and EGF increased myocardial superoxide production (187 ± 9xa0%, 149 ± 8xa0% and 163.7 ± 6xa0% of control, respectively) and AG1478 inhibited these effects. Interestingly, Los inhibited only Ang II whilst BQ123 cancelled both Ang II and ET-1 actions, supporting the sequential and unidirectional activation of AT1, ETA and EGFR. Based on the present evidence, we propose that endogenous ET-1 mediates the hypertrophic response to Ang II by a mechanism that involves EGFR transactivation and redox-dependent activation of the ERK1/2-p90(RSK) and NHE-1 in adult cardiomyocytes.