EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Quantification of carbamazepine and its 10,11-epoxide metabolite in rat plasma by UPLC-UV and application to pharmacokinetic study.
PMID 24327551
Abstract
A rapid, selective and sensitive UPLC-UV method was developed and validated for the quantitative analysis of carbamazepine and its epoxide metabolite in rat plasma. A relatively small volume of plasma sample (200 μL) is required for the described analytical method. The method includes simple protein precipitation, liquid-liquid extraction, evaporation, and reconstitution steps. Samples were separated on a Waters Acquity UPLC BEH C18 column (1.7 µm, 2.1 × 100 mm) with a gradient mobile phase consisted of 60:40 going to 40:60 (v/v) water-acetonitrile at a flow rate of 0.5 mL/min. The total run time was as low as 6 min, representing a significant improvement in comparison to existing methods. Excellent linearity (r(2)  > 0.999) was achieved over a wide concentration range. Close to complete recovery, short analysis time, high stability, accuracy, precision and reproducibility, and low limit of quantitation were demonstrated. Finally, we successfully applied this analytical method to a pre-clinical oral pharmacokinetic study, revealing the plasma profiles of both carbamazepine and carbamazepine-10,11-epoxide following oral administration of carbamazepine to rats. The advantages demonstrated in this work make this analytical method both time- and cost-efficient approach for drug and metabolite monitoring in the pre-clinical/clinical laboratory.