EMAIL THIS PAGE TO A FRIEND
International journal of endocrinology
Investigation of responsiveness to thyrotropin-releasing hormone in growth hormone-producing pituitary adenomas.
PMID 24348552
Abstract
Objective. The aim of this study was to investigate how the paradoxical response of GH secretion to TRH changes according to tumor volumes. Methods. Patients with newly diagnosed acromegaly were classified as either TRH responders or nonresponders according to the results of a TRH stimulation test (TST), and their clinical characteristics were compared according to responsiveness to TRH and tumor volumes. Results. A total of 41 acromegalic patients who underwent the TST were included in this study. Between TRH responders and nonresponders, basal GH, IGF-I levels, peak GH levels, and tumor volume were not significantly different, but the between-group difference of GH levels remained near significant over the entire TST time. ΔGHmax-min during the TST were significantly different according to the responsiveness to TRH. Peak GH levels and ΔGHmax-min during the TST showed significantly positive correlations with tumor volume with higher levels in macroadenomas than in microadenomas. GH levels over the entire TST time also remained significantly higher in macroadenomas than in microadenomas. Conclusion. Our data demonstrated that the paradoxical response of GH secretion to TRH in GH-producing pituitary adenomas was not inversely correlated with tumor volumes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O1014
Octreotide, ≥98% (HPLC)
C49H66N10O10S2