EMAIL THIS PAGE TO A FRIEND
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology
Comparison between effects of myo-inositol and D-chiro-inositol on ovarian function and metabolic factors in women with PCOS.
PMID 24351072
Abstract
Myo-inositol and D-chiro-inositol are capable of improving the ovarian function and metabolism of polycystic ovary syndrome (PCOS) patients. The aim of this work is to compare the effects of myo-inositol and D-chiro-inositol in PCOS. We enrolled 50 patients, with homogeneous bio-physical features, affected by PCOS and menstrual irregularities, and we randomly divided them into two groups: 25 were treated with 4 g of myo-inositol/die plus 400 mcg of folic acid/die orally for six months, 25 with 1 g of D-chiro-inositol/die plus 400 mcg of folic acid/die orally for six months. We analyzed in both groups pre-treatment and post-treatment BMI, systolic and diastolic blood pressure, Ferriman-Gallwey score, Cremoncini score, serum LH, LH/FSH ratio, total and free testosterone, dehydroepiandrosterone sulfate (DHEA-S), Δ-4-androstenedione, SHBG, prolactin, glucose/immunoreactive insulin (IRI) ratio, homeostatic model assessment (HOMA) index, and the resumption of regular menstrual cycles. Both the isoforms of inositol were effective in improving ovarian function and metabolism in patients with PCOS, although myo-inositol showed the most marked effect on the metabolic profile, whereas D-chiro-inositol reduced hyperandrogenism better.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74137
D-chiro-Inositol, ≥98.0% (HPLC)
C6H12O6