EMAIL THIS PAGE TO A FRIEND
F1000Research
Rapid doubling of Alzheimer's amyloid-β40 and 42 levels in brains of mice exposed to a nickel nanoparticle model of air pollution.
PMID 24358824
Abstract
Over 20 genetic risk factors have been confirmed to associate with elevated risk for Alzheimer's disease (AD), but the identification of environmental and/or acquired risk factors has been more elusive. At present, recognized acquired risks for AD include traumatic brain injury, hypercholesterolemia, obesity, hypertension, and type 2 diabetes. Based on reports associating various inhalants with AD pathology, we investigated the possibility that air pollution might contribute to AD risk by exposing wild-type mice to a standard air pollution modeling system employing nickel nanoparticle-enriched atmosphere for 3 hr. Mice exposed to air pollution showed 72-129% increases in brain levels of both amyloid-β peptides Aβ40 and Aβ42, as well as Aβ42/40 (p <0.01). These effects on elevation of brain Aβ exceed those associated with trisomy 21, a known risk for early onset AD pathology, raising the possibility that clinical importance might be attached. Further work is required to establish the molecular and physiological basis for these phenomena. The rapid, dramatic effect, if verified, would suggest that inhalant exposures should be evaluated for their possible roles in contributing to the environmental risk for common forms of AD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
283622
Nickel(II) hydroxide
H2NiO2