EMAIL THIS PAGE TO A FRIEND
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society
Checkpoint kinase inhibitor AZD7762 overcomes cisplatin resistance in clear cell carcinoma of the ovary.
PMID 24362713
Abstract
Checkpoint kinase (Chk) inhibitors are thought to increase the cytotoxic effects of DNA-damaging agents and are undergoing clinical trials. The present study was aimed to assess the potential to use the Chk1 and Chk2 inhibitor, AZD7762, with other anticancer agents in chemotherapy to treat ovarian clear cell carcinoma. Four ovarian clear cell carcinoma cell lines were used in this study. We treated the cells with AZD7762 and anticancer agents, then assessed cell viability, cell cycle distribution, apoptosis, and the expression of protein in apoptotic pathways and molecules downstream of the Chk signaling pathways. We also investigated the effects of these drug combinations on tumor growth in a nude mouse xenograft model. Synergistic effects from the combination of AZD7762 and cisplatin were observed in all 4 cell lines. However, we observed additive effects when AZD7762 was combined with paclitaxel on all cell lines tested. AZD7762 effectively suppressed the Chk signaling pathways activated by cisplatin, dramatically enhanced expression of phosphorylated H2A.X, cleaved caspase 9 and PARP, decreased the proportion of cells in the gap 0/ gap 1 phase and the synthesis-phase fraction, and increased apoptotic cells. Combinations of small interfering RNA against Chk 1 and small interfering RNA against Chk2 enhanced the cytotoxic effect of cisplatin in both RMG-I and KK cells. Finally, treating mice-bearing RMG-I with AZD7762 and cisplatin significantly suppressed growth of tumors in a xenograft model. The present study indicates that chemotherapy with AZD7762 and cisplatin should be explored as a treatment modality for women with ovarian clear cell carcinoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA001473
Anti-CASP9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503332
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503333
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503334
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503335
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
C7729 Anti-Caspase 9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300683
Anti-Caspase 9 antibody produced in rabbit, affinity isolated antibody
SAB3500405
Anti-Caspase-9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001878
Anti-CHEK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV30290
Anti-CHEK2 antibody produced in rabbit, affinity isolated antibody
SAB2100415
Anti-CHEK2 antibody produced in rabbit, affinity isolated antibody
SAB3500192
Anti-Chk2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300514
Anti-H2AFX (Ab-139) antibody produced in rabbit, affinity isolated antibody
PLA0294
Goat anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
61506
L-Threonine, certified reference material, TraceCERT®
C4H9NO3
PHR1242
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
C4H9NO3
T1340000
L-Threonine, European Pharmacopoeia (EP) Reference Standard
C4H9NO3
WH0000993M1
Monoclonal Anti-CDC25A antibody produced in mouse, clone 3D5, purified immunoglobulin, buffered aqueous solution
WH0001111M5
Monoclonal Anti-CHEK1 antibody produced in mouse, clone 2G3, purified immunoglobulin, buffered aqueous solution
SAB5300524 Monoclonal Anti-CHEK1 antibody produced in mouse, clone 2G1D5, ascites fluid
C9358
Monoclonal Anti-Chk1 antibody produced in mouse, ~2 mg/mL, clone DCS-310, purified immunoglobulin, buffered aqueous solution
WH0003014M5 Monoclonal Anti-H2AFX antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
SAB1403902 Monoclonal Anti-H2AFX, (N-terminal) antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
Y0000698
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
C47H51NO14
Y0000700
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard
C47H51NO14
Y0000719
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
C47H51NO14
Y0000739
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
C47H51NO14
PLA0122
Rabbit anti-Chk2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0023
Rabbit anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.