EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
LC-MS/MS analysis of uncommon paracetamol metabolites derived through in vitro polymerization and nitration reactions in liquid nitrogen.
PMID 24365200
Abstract
Paracetamol (acetaminophen, APAP) is a commonly used analgesic drug. Known paracetamol metabolites include the glucuronide, sulfate and mercapturate. N-Acetyl-benzoquinonimine (NAPQI) is considered the toxic intermediate metabolite of paracetamol. In vitro and in vivo studies indicate that paracetamol is also metabolized to additional poorly characterized metabolites. For example, metabolomic studies in urine samples of APAP-treated mice revealed metabolites such as APAP-sulfate-APAP and APAP-S-S-APAP in addition to the classical phase II metabolites. Here, we report on the development and application of LC-MS and LC-MS/MS approaches to study reactions of unlabelled and (2)H-labelled APAP with unlabelled and (15)N-labelled nitrite in aqueous phosphate buffers (pH 7.4) upon their immersion into liquid nitrogen (-196°C). In mechanistic studies, these reactions were also studied in aqueous buffer prepared in (18)O-labelled water. LC-MS and LC-MS/MS analyses were performed on a reverse-phase material (C18) using gradient elution (2mM ammonium acetate/acetonitrile), in positive and negative electrospray mode. We identified a series of APAP metabolites including di-, tri- and tetra-APAP, mono- and di-nitro-APAP and nitric ester of di-APAP. Our study indicates that nitrite induces oxidation, i.e., polymerization and nitration of APAP, when buffered APAP/nitrite solutions are immersed into liquid nitrogen. These reactions are specific for nitrite with respect to nitrate and do not proceed via intermediate formation of NAPQI. Potassium ions and physiological saline but not thiols inhibit nitrite- and shock-freeze-induced reactions of paracetamol. The underlying mechanism likely involves in situ formation of NO2 radicals from nitrite secondary to profound pH reduction (down to pH 1) and disproportionation. Polymeric paracetamol species can be analyzed as pentafluorobenzyl derivatives by LC-MS but not by GC-MS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
101052
2,3,4,5,6-Pentafluorobenzyl bromide, 99%
C7H2BrF5
C3290000
Cysteine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard
C3H7NO2S · HCl · H2O
C121800
L-Cysteine hydrochloride hydrate, 99%
C3H7NO2S · HCl · xH2O
C7880
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
C3H7NO2S · HCl · H2O
C6852
L-Cysteine hydrochloride monohydrate, from non-animal source, cell culture tested, meets EP, USP testing specifications
C3H7NO2S · HCl · H2O
C5610
L-Cysteine hydrochloride monohydrate, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2S · HCl · H2O
30122
L-Cysteine hydrochloride monohydrate, tested according to Ph.Eur.
C3H7NO2S · HCl · H2O
W779989
L-Cysteine hydrochloride monohydrate, FG, produced by Wacker Chemie AG, Burghausen, Germany, Food, 20 mesh
C3H7NO2S · HCl · H2O
W779865
L-Cysteine hydrochloride monohydrate, 99%, FG, produced by Wacker Chemie AG, Burghausen, Germany
C3H7NO2S · HCl · H2O
PHR1102
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO2S · HCl · H2O
30129
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
C3H7NO2S · HCl · H2O
30130
L-Cysteine hydrochloride monohydrate, ≥99.0% (RT)
C3H7NO2S · HCl · H2O
778451
L-Cysteine hydrochloride monohydrate, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.5-101.0% (Ph.Eur (2.2.20))
C3H7NO2S · HCl · H2O
33001
pentafluorobenzyl bromide (PFB-Br), analytical standard
C7H2BrF5
15736
Sodium nitrate, analytical standard
NNaO3
347663
Sodium nitrate, beads, 1-2 mm, 98%
NNaO3