EMAIL THIS PAGE TO A FRIEND
Acta histochemica
Epigallocatechin-3-gallate (EGCG) attenuates concanavalin A-induced hepatic injury in mice.
PMID 24373695
Abstract
(-)-Epigallocatechin-3-gallate (EGCG) is the most abundant polyphenolic compound present in green tea and has been shown to possess anti-inflammatory and anti-oxidative properties. In this study, we investigated the protective effects of EGCG against concanavalin A (ConA)-induced liver injury and the underlying mechanisms. EGCG (5 mg/kg) was administered orally by gavage to mice twice daily for 10 days before an intravenous injection of ConA. We found that EGCG effectively rescued lethality, improved hepatic pathological damage, and decreased serum levels of alanine aminotransaminase (ALT) in ConA-challenged mice. Furthermore, EGCG also significantly prevented the release of tumor necrosis factor (TNF)-α, interferon (IFN)-γ, interleukin (IL)-4, and IL-6 in serum, reduced malondialdehyde (MDA) levels, and restored glutathione (GSH) content and superoxide dismutase (SOD) activity in liver tissues from ConA-challenged mice. Finally, nuclear factor (NF)-κB activation and expression levels of Toll-like receptor (TLR) 2, TLR4 and TLR9 protein in liver tissues were significantly inhibited by EGCG pretreatment. Taken together, our data suggest that EGCG possesses hepatoprotective properties against ConA-induced liver injury through its anti-inflammatory and anti-oxidant actions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22787 Anti-TLR2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
PRS3135
Anti-TLR2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
WH0007097M7
Monoclonal Anti-TLR2 antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution
SAB1404474
Monoclonal Anti-TLR2 antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
SAB1409745
Monoclonal Anti-TLR2 antibody produced in mouse, clone 2A3, purified immunoglobulin, buffered aqueous solution
SAB1409746 Monoclonal Anti-TLR2 antibody produced in mouse, clone 2D2, purified immunoglobulin, buffered aqueous solution
SAB1409747 Monoclonal Anti-TLR2 antibody produced in mouse, clone 3F10, purified immunoglobulin, buffered aqueous solution
SAB1409748 Monoclonal Anti-TLR2 antibody produced in mouse, clone 1E3, purified immunoglobulin, buffered aqueous solution
SAB1409749 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4D6, purified immunoglobulin, buffered aqueous solution
SAB1409750 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4G9, purified immunoglobulin, buffered aqueous solution
SAB1409751 Monoclonal Anti-TLR2 antibody produced in mouse, clone 1C6, purified immunoglobulin, buffered aqueous solution
SAB1409752 Monoclonal Anti-TLR2 antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution
SAB1409753 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4C4, purified immunoglobulin, buffered aqueous solution
SAB1409754 Monoclonal Anti-TLR2 antibody produced in mouse, clone 3H8, purified immunoglobulin, buffered aqueous solution
SAB1409755 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4H6, purified immunoglobulin, buffered aqueous solution