EMAIL THIS PAGE TO A FRIEND
International journal of oncology
The CDK1 inhibitor RO3306 improves the response of BRCA-proficient breast cancer cells to PARP inhibition.
PMID 24378347
Abstract
Breast cancer is one of the most common malignancies in women. Approximately 15% of the patients belong to the triple-negative breast cancer (TNBC) group, and have the disadvantage of not benefiting from currently available receptor-targeted systemic therapies. Some cancers in the TNBC group harbor defects in DNA double-strand break repair by homologous recombination (HR), such as BRCA1 dysfunction, and are hypersensitive to poly (ADP-ribose) polymerase (PARP) inhibition. However, only a small fraction of the tumors are BRCA-deficient, and this restricts the therapeutic utility of the PARP inhibitor monotherapy. Cyclin-dependent kinasexa01 (CDK1) is necessary not only for BRCA1-mediated Sxa0phase checkpoint activation, but also for HR, because it phosphorylates BRCA1 for the efficient formation of BRCA1 foci. In this study, we showed that the combined inhibition of CDK1 and PARP in BRCA-proficient MDA-MB-231 breast cancer cells resulted in dramatically reduced cell growth compared to PARP inhibition alone. Mechanistic investigations revealed that this sensitivity appears to be mediated by sustained DNA damage and inefficient DNA repair triggering mitochondrial-mediated apoptosis as well as autophagy. Our results suggest that CDK1 inhibition represents a plausible strategy for expanding the utility of PARP inhibitors to BRCA‑proficient breast cancers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21183 Anti-BRCA1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV02017
Anti-BRCA1 antibody produced in rabbit, affinity isolated antibody
SAB2104043
Anti-BRCA1 antibody produced in rabbit, affinity isolated antibody
HPA034966 Anti-BRCA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503332
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503333
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503334
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503335
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4300683
Anti-Caspase 9 antibody produced in rabbit, affinity isolated antibody
SAB3500405
Anti-Caspase-9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100378
Anti-CDC2 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100379
Anti-CDC2 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2103891
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
SAB4500051
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
SAB4503707
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
HPA003387
Anti-CDK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300348
Anti-PGR (Ab-190) antibody produced in rabbit, affinity isolated antibody
HPA004751
Anti-PGR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV100687
Anti-PGR antibody produced in rabbit, affinity isolated antibody
SAB2101781
Anti-PGR antibody produced in rabbit, affinity isolated antibody
HPA008428
Anti-PGR antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105280
Anti-PGR antibody produced in rabbit, affinity isolated antibody
HPA017176
Anti-PGR antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22679 Anti-Progesterone Receptor antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4502184
Anti-Progesterone Receptor antibody produced in rabbit, affinity isolated antibody
SAB4502185
Anti-Progesterone Receptor antibody produced in rabbit, affinity isolated antibody
SAB4502186
Anti-Progesterone Receptor antibody produced in rabbit, affinity isolated antibody
AV33244
Anti-RORC (AB1) antibody produced in rabbit, affinity isolated antibody
AV45622
Anti-RORC (AB2) antibody produced in rabbit, affinity isolated antibody
SAB2501680
Anti-RORC antibody produced in goat, affinity isolated antibody, buffered aqueous solution