EMAIL THIS PAGE TO A FRIEND
TheScientificWorldJournal
Speciation studies of diorganotin(IV) complexes with 3,3-bis(1-methylimidazol-2-yl)propionate--displacement reaction by DNA constituents.
PMID 24385867
Abstract
The interaction of 3,3-bis(1-methylimidazol-2-yl)propionate (BIMP) with dimethyltin(IV) dichloride (DMT), dibutyltin(IV) dichloride (DBT), and diphenyltin(IV) dichloride (DPT) is investigated at 25°C and 0.1 M ionic strength in water for dimethyltin(IV), and in a 50% dioxane-water mixture for dibutyltin(IV) and diphenyltin(IV). The stepwise formation constants of the 1 : 1 and 1 : 2 complexes formed in solution are calculated from potentiometric measurements using the nonlinear least-square program MINIQUAD-75. The concentration distribution of the various complex species is evaluated as a function of pH. Displacement reactions of the coordinated 3,3-bis(1-methylimidazol-2-yl)propionate by inosine and inosine-5'-monophosphate are investigated from calculations based upon equilibrium properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205494
Dibutyltin dichloride, 96%
C8H18Cl2Sn
229202
Diphenyltin dichloride, 96%
C12H10Cl2Sn