EMAIL THIS PAGE TO A FRIEND
Pflugers Archiv : European journal of physiology
microRNA expression and its potential role in cardioprotection by ischemic postconditioning in pigs.
PMID 24390754
Abstract
Ischemic postconditioning (PoCo) reduces infarct size following myocardial ischemia/reperfusion. To protect, PoCo must be performed early during reperfusion, and causal cardioprotective signaling must occur then. The role of microRNA (miRNA) in PoCo is unclear. Anesthetized pigs were subjected to 60 min left anterior descending coronary artery (LAD) occlusion and 180 min reperfusion. Immediate full reperfusion (IFR, n = 5) was compared to PoCo (four cycles of 60 s/60 s reperfusion/reocclusion, n = 5). Transmural myocardial biopsies from the LAD territory were sampled at baseline, 60 min ischemia, 10 and 180 min reperfusion. RNA was isolated. The expression of 11 miRNAs, including muscle-specific (miRNA-1, -133a, -206, -208b, -214, and -499), fibrosis- (miRNA-21, -24, and -29b), neovascularization- (miRNA-92a), and inflammation-associated (miRNA-146b) candidates, was quantified using real-time PCR (RT-PCR). mRNA expression at baseline and 180 min reperfusion was quantified and validated (microarray and RT-PCR). PoCo reduced infarct size from 44.9 ± 7.7 to 34.8 ± 5.3% of the area at risk. The expression of miRNA-1, -24, -29b, -133a, -146b, -208b, and -499 was increased at 10 min reperfusion with PoCo vs. IFR; however, that of miRNA-1, -24, -208b, and -499 was already increased at 60 min ischemia and probably reflects falsely positive results. Five mRNAs were different with PoCo vs. IFR. In silico analysis identified a tentative connection between three miRNAs and five mRNAs with the biological functions "cell death", "inflammatory response" and/or "glucose metabolism". If at all, only miRNA-29b, -133a, and -146b fulfill the minimal temporal requirements for a potential causal involvement in cardioprotection by PoCo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E6530
Etomidate, >98% (HPLC), powder
C14H16N2O2