EMAIL THIS PAGE TO A FRIEND
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
Circulating TSLP associates with decreased wheezing in non-atopic preschool children: data from the URECA birth cohort.
PMID 24397611
Abstract
Mouse models of atopic march suggest that systemic, skin-derived thymic stromal lymphopoietin (TSLP) mediates progression from eczema to asthma. We investigated whether circulating TSLP is associated with eczema, allergic sensitization, or recurrent wheezing in young children. A prospective analysis of the relationship between plasma levels of TSLP to allergic sensitization and recurrent wheezing was conducted in the birth cohort from the Urban Environment and Childhood Asthma (URECA) study. Plasma TSLP levels were measured at 1, 2, and 3xa0years of age and analysed for correlation with clinical parameters in each of the three years. Only those children with consecutive samples for all three years were included in this analysis. We detected TSLP in 33% of 236 children for whom plasma samples were available for all three years. Overall, a consistently significant association was not found between TSLP and eczema or allergic sensitization. With regard to recurrent wheezing, children with detectable TSLP at one year of age were significantly less likely to experience recurrent wheezing by 3xa0years compared with those children without detectable TSLP, but this was only seen in children without aeroallergen sensitization at 3xa0years (Pxa0