EMAIL THIS PAGE TO A FRIEND
Mucosal immunology
Neutrophil infiltration favors colitis-associated tumorigenesis by activating the interleukin-1 (IL-1)/IL-6 axis.
PMID 24424523
Abstract
Neutrophil infiltration is a key event in chronic intestinal inflammation and associated colorectal cancer, but how these cells support cancer development is poorly understood. In this study, using a mouse model of colitis-associated cancer (CAC), we have demonstrated that infiltrated neutrophils produce large amounts of interleukin-1 (IL)-1β that is critical for the development of CAC. Depletion of neutrophil or blockade of IL-1β activity significantly reduced mucosal damage and tumor formation. This protumorigenic function of IL-1β was mainly attributed to increased IL-6 secretion by intestine-resident mononuclear phagocytes (MPs). Furthermore, commensal flora-derived lipopolysaccharide (LPS) was identified to trigger IL-1β expression in neutrophils. Importantly, accumulation of IL-1β-expressing neutrophils was seen in lesions of patients suffering from ulceratic CAC and these infiltrated neutrophils induced IL-6 production by intestinal MPs in an IL-1β-dependent manner. Overall, these findings reveal that in CAC milieu, infiltrating neutrophils secrete IL-1β that promotes tumorigenesis by inducing IL-6 production by intestinal MPs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5486
Azoxymethane, 13.4 M, ≥98%
C2H6N2O