EMAIL THIS PAGE TO A FRIEND
Cerebral cortex (New York, N.Y. : 1991)
Transient Maternal Hypothyroxinemia Potentiates the Transcriptional Response to Exogenous Thyroid Hormone in the Fetal Cerebral Cortex Before the Onset of Fetal Thyroid Function: A Messenger and MicroRNA Profiling Study.
PMID 24436321
Abstract
Thyroid hormone (TH) is essential for brain development both before and after birth. We have used gene expression microarrays to identify TH-regulated genes in the fetal cerebral cortex prior to the onset of fetal thyroid function to better understand the role of TH in early cortical development. TH levels were transiently manipulated in pregnant mice by treatment with goitrogens from gestational day (GD) 13-16 and/or by injection of TH 12 h before sacrifice on GD 16. The transcriptional response to exogenous TH in the GD 16 fetal cortex was potentiated by transient goitrogen treatment, suggesting that the hypothyroxinemic brain is a different substrate upon which TH can act, or that robust compensatory mechanisms are induced by transient hypothyroxinemia. Several known TH-responsive genes were identified including Klf9, and several novel TH-responsive genes such as Appbp2, Ppap2b, and Fgfr1op2 were identified in which TH response elements were confirmed. We also identified specific microRNAs whose expression in the fetal cortex was affected by TH treatment, and determined that Ppap2b and Klf9 are the target genes of miR-16 and miR-106, respectively. Thus, a complex redundant functional network appears to coordinate TH-mediated gene expression in the developing brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306490
ANTI-CD71(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501233
Anti-F2R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405778
Anti-F2R antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100740
Anti-F2R antibody produced in rabbit, affinity isolated antibody
R8656
Anti-Peroxiredoxin 2 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2500777
Anti-Peroxiredoxin 2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
GW21018 Anti-Peroxiredoxin-2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406520
Anti-PRDX2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101878
Anti-PRDX2 antibody produced in rabbit, affinity isolated antibody
GW23058 Anti-TERT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2502024
Anti-TERT antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501035
Anti-Thioredoxin Reductase 1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T5700 Anti-Thrombin Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA015587
Anti-TMEFF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500131
Anti-TRXR2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1400287
Anti-TXNRD1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA001395
Anti-TXNRD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411372
Anti-TXNRD1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA003323
Anti-TXNRD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2702064
Anti-TXNRD2 antibody produced in rabbit
SAB1402186
Monoclonal Anti-F2R antibody produced in mouse, clone 2C5, purified immunoglobulin, buffered aqueous solution
WH0007001M1
Monoclonal Anti-PRDX2 antibody produced in mouse, clone 4E10-2D2, purified immunoglobulin, buffered aqueous solution
SAB1404456
Monoclonal Anti-PRDX2 antibody produced in mouse, clone S2, purified immunoglobulin, buffered aqueous solution
SAB1403162
Monoclonal Anti-TMEFF2 antibody produced in mouse, clone 1D12, purified immunoglobulin, buffered aqueous solution
SAB4100469
Monoclonal Anti-TMEFF2 antibody produced in mouse, clone TMEFF2-A406, culture supernatant
WH0010587M1 Monoclonal Anti-TXNRD2 antibody produced in mouse, clone 3A7-F1, purified immunoglobulin, buffered aqueous solution
SRP6081
TXNRD1 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE)