EMAIL THIS PAGE TO A FRIEND
Electroanalysis
The Application of Nafion Metal Catalyst Free Carbon Nanotube Modified Gold Electrode: Voltammetric Zinc Detection in Serum.
PMID 24436574
Abstract
Metal catalyst free carbon nanotube (MCFCNT) whiskers were first used as an electrode modification material on a gold electrode surface for zinc voltammetric measurements. A composite film of Nafion and MCFCNT whiskers was applied to a gold electrode surface to form a mechanically stable sensor. The sensor was then used for zinc detection in both acetate buffer solution and extracted bovine serum solution. A limit of detection of 53 nM was achieved for a 120 s deposition time. The zinc in bovine serum was extracted via a double extraction procedure using dithizone in chloroform as a zinc chelating ligand. The modified electrode was found to be both reliable and sensitive for zinc measurements in both matrices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194832
Dithizone, ACS reagent, ≥85.0%
C13H12N4S