EMAIL THIS PAGE TO A FRIEND
The Journal of arthroplasty
Pulse lavage is inadequate at removal of biofilm from the surface of total knee arthroplasty materials.
PMID 24439797
Abstract
In acute periprosthetic infection, irrigation and debridement with component retention has a high failure rate in some studies. We hypothesize that pulse lavage irrigation is ineffective at removing biofilm from total knee arthroplasty (TKA) components. Staphylococcus aureus biofilm mass and location was directly visualized on arthroplasty materials with a photon collection camera and laser scanning confocal microscopy. There was a substantial reduction in biofilm signal intensity, but the reduction was less than a ten-fold decrease. This suggests that irrigation needs to be further improved for the removal of biofilm mass below the necessary bioburden level to prevent recurrence of acute infection in total knee arthroplasty.