EMAIL THIS PAGE TO A FRIEND
The International journal of oral & maxillofacial implants
Ultraviolet photofunctionalization of titanium implants.
PMID 24451893
Abstract
In the face of growing demands and challenges in implant therapy, implant surfaces with improved biologic capabilities are required. This review paper summarizes the findings of recent in vitro and in vivo studies related to ultraviolet (UV) photofunctionalization of titanium. UV photofunctionalization is defined as an overall phenomenon of modification of titanium surfaces occuring after UV treatment, including the alteration of physicochemical properties and the enhancement of biologic capabilities. Bone morphogenesis around UV-treated titanium implants is distinctly improved compared with that seen around untreated control implants, leading to rapid and complete establishment of osseointegration with nearly 100% bone-to-implant contact in an animal model, as opposed to less than 55% for untreated implants. A series of in vitro studies demonstrated considerable enhancement of attachment, retention, and subsequent functional cascades of osteogenic cells derived from animals and humans after UV treatment. UV treatment converts titanium surfaces from hydrophobic to superhydrophilic and removes unavoidably contaminated hydrocarbons. UV-treated titanium surfaces also manifest a unique electrostatic status and act as direct cell attractants without the aid of ionic and organic bridges, which imparts a novel physicochemical functionality to titanium, which has long been understood as a bioinert material. UV treatment is simple and low in cost, and it has been proven effective for all types of titanium surfaces tested. These data suggest that UV photofunctionalization can be a novel, effective measure to improve implant therapy in the dental and orthopedic fields. Future research will focus on validating these findings in clinical studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
IRMM531A
Titanium, IRMM® certified Reference Material, 0.1 mm foil
Ti
IRMM531B
Titanium, IRMM® certified Reference Material, 0.5 mm foil
Ti
IRMM531C
Titanium, IRMM® certified Reference Material, 0.5 mm wire
Ti
305812
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Ti
433667
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Ti
369489
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Ti
267503
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Ti
460397
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Ti
348791
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Ti
348821
Titanium, foil, thickness 0.05 mm, 99.99% trace metals basis
Ti
268496
Titanium, powder, −100 mesh, 99.7% trace metals basis
Ti
366994
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Ti
513415
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Ti
347132
Titanium, rod, diam. 6.35 mm, 99.99% trace metals basis
Ti
266051
Titanium, rod, diam. 6.35 mm, 99.7% trace metals basis
Ti
268526
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Ti
348856
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Ti
266035
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Ti
267902
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Ti
348864
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Ti
460400
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Ti
266019
Titanium, wire, diam. 0.127 mm, 99.99% trace metals basis
Ti
767506
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Ti
GF57414179
Titanium, rod, 1000mm, diameter 6mm, annealed, 99.6+%
Ti
GF72693883
Titanium, rod, 100mm, diameter 1.5mm, annealed, 99.6+%
Ti
GF76772391
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Ti
GF39040084
Titanium, rod, 100mm, diameter 16mm, annealed, 99.6+%
Ti
GF87716997
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Ti
GF35446316
Titanium, rod, 100mm, diameter 20mm, annealed, 99.6+%
Ti
GF09714719
Titanium, rod, 100mm, diameter 20mm, as drawn, 99.99+%
Ti