EMAIL THIS PAGE TO A FRIEND
Angiology
The effect of adding ezetimibe to rosuvastatin on renal function in patients undergoing elective vascular surgery.
PMID 24458801
Abstract
We compared the effects of lipid lowering with rosuvastatin (RSV) monotherapy versus intensified treatment by combining RSV with ezetimibe (EZT) on kidney function in patients undergoing vascular surgery. Patients were randomly assigned to either 10 mg/d RSV (n = 136) or RSV 10 mg/d plus EZT 10 mg/d (RSV/EZT, n = 126). At 12 months, a similar decrease in estimated glomerular filtration rate (eGFR) was noted. Patients who achieved a low-density lipoprotein cholesterol (LDL-C) of <100 mg/dL had less eGFR decrease than those patients having an LDL-C limit of more than 100 mg/dL. There were no significant changes in the urinary total protein to creatinine ratio in either group. Significant microalbuminuria was evident in both the groups. Patients undergoing vascular surgery show deterioration in their renal function during the first year, despite statin therapy. Intensified lipid-lowering therapy by adding EZT does not appear to have any renoprotective effect.