EMAIL THIS PAGE TO A FRIEND
Acta diabetologica
Glycogen storage in the human retinal pigment epithelium: a comparative study of diabetic and non-diabetic donors.
PMID 24458975
Abstract
Liver and muscle glycogen content is reduced in diabetic patients but there is no information on the effect of diabetes on the glycogen content in the retinal pigment epithelium (RPE). The main aim of the study was to compare the glycogen content in the RPE between diabetic and non-diabetic human donors. Glycogen synthase (GS) and glycogen phosphorylase (GP), the key enzymes of glycogen metabolism, as well as their isoforms, were also assessed. For this purpose, 44 human postmortem eye cups were included (22 from 11 type 2 diabetic and 22 from 11 non-diabetic donors matched by age). Human RPE cells cultured in normoglycemic and hyperglycemic conditions were also analyzed. Glycogen content as well as the mRNA, protein content and enzyme activity of GS and GP were determined. In addition, GS and GP isoforms were characterized. In the RPE from diabetic donors, as well as in RPE cells grown in hyperglycemic conditions, the glycogen content was increased. The increase in glycogen content was associated with an increase in GS without changes in GP levels. In RPE form human donors, the muscle GS isoform but not the liver GS isoform was detected. Regarding GP, the muscle and brain isoform of GP but not the liver GP isoform were detected. We conclude that glycogen storage is increased in the RPE of diabetic patients, and it is associated with an increase in GS activity. Further studies aimed at determining the role of glycogen deposits in the pathogenesis of diabetic retinopathy are warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4200176
Anti-JNK antibody, Mouse monoclonal, clone 1C2, purified from hybridoma cell culture
SAB4502398
Anti-JNK1 antibody produced in rabbit, affinity isolated antibody
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
SAB1405665
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
SAB1406300
Anti-MAPK8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
M0800
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
J2253 Anti-phospho-c-Jun (pSer73) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504369
Anti-phospho-c-Jun (pSer73) antibody produced in rabbit, affinity isolated antibody
SAB4300002
Anti-phospho-JUN (pSer73) antibody produced in rabbit, affinity isolated antibody
P1491 Anti-phospho-p38 (pThr180/pTyr182) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
GW21175B Anti-RK antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
M7431 Monoclonal Anti-MAP Kinase 2 (ERK-2) antibody produced in mouse, clone 1B3B9, purified immunoglobulin, buffered aqueous solution
WH0005599M2
Monoclonal Anti-MAPK8 antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
SAB1404249
Monoclonal Anti-MAPK8 antibody produced in mouse, clone 2F11, purified immunoglobulin, buffered aqueous solution