EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Heterocycles 33: lipophilicity of a new class of thioethers estimated by reversed-phase thin-layer chromatography and different computational methods.
PMID 24464959
Abstract
The retention behavior for a series of new polyheterocyclic compounds containing azolic rings (1,3-thiazole, 1,2,4-triazole, 1,3,4-oxadiazole, 1,3,4-thiadiazole) has been investigated using reversed-phase thin-layer chromatography. Different approaches and computational methods were employed to evaluate their lipophilicity indices derived from chromatographic parameters. The obtained experimental results were correlated with various lipophilicity indices estimated via different computer software and internet websites. A strong correlation between experimental and computed results was observed. Furthermore, the lipophilicity parameters obtained by applying principal component analysis divided the investigated compounds into four groups according to their structural similarities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T46108
1,2,4-Triazole, 98%
C2H3N3