PloS one
Familial young-onset diabetes, pre-diabetes and cardiovascular disease are associated with genetic variants of DACH1 in Chinese.
PMID 24465431
Abstract
In Asia, young-onset type 2 diabetes (YOD) is characterized by obesity and increased risk for cardiovascular disease (CVD). In a genome-wide association study (GWAS) of 99 Chinese obese subjects with familial YOD diagnosed before 40-year-old and 101 controls, the T allele of rs1408888 in intron 1 of DACH1(Dachshund homolog 1) was associated with an odds ratio (OR) of 2.49(95% confidence intervals:1.57-3.96, P = 8.4 × 10(-5)). Amongst these subjects, we found reduced expression of DACH1 in peripheral blood mononuclear cells (PBMC) from 63 cases compared to 65 controls (P = 0.02). In a random cohort of 1468 cases and 1485 controls, amongst top 19 SNPs from GWAS, rs1408888 was associated with type 2 diabetes with a global P value of 0.0176 and confirmation in a multiethnic Asian case-control cohort (7370/7802) with an OR of 1.07(1.02-1.12, P(meta)  = 0.012). In 599 Chinese non-diabetic subjects, rs1408888 was linearly associated with systolic blood pressure and insulin resistance. In a case-control cohort (n = 953/953), rs1408888 was associated with an OR of 1.54(1.07-2.22, P = 0.019) for CVD in type 2 diabetes. In an autopsy series of 173 non-diabetic cases, TT genotype of rs1408888 was associated with an OR of 3.31(1.19-9.19, P = 0.0214) and 3.27(1.25-11.07, P = 0.0184) for coronary heart disease (CHD) and coronary arteriosclerosis. Bioinformatics analysis revealed that rs1408888 lies within regulatory elements of DACH1 implicated in islet development and insulin secretion. The T allele of rs1408888 of DACH1 was associated with YOD, prediabetes and CVD in Chinese.