EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Topical photodynamic therapy following excisional wounding of human skin increases production of transforming growth factor-β3 and matrix metalloproteinases 1 and 9, with associated improvement in dermal matrix organization.
PMID 24471979
Abstract
Animal studies report photodynamic therapy (PDT) to improve healing of excisional wounds; the mechanism is uncertain and equivalent human studies are lacking. To explore the impact of methyl aminolaevulinate (MAL)-PDT on clinical and microscopic parameters of human cutaneous excisional wound healing, examining potential modulation through production of transforming growth factor (TGF)-β isoforms. In 27 healthy older men (60-77 years), a 4-mm punch biopsy wound was created in skin of the upper inner arm and treated with MAL-PDT three times over 5 days. An identical control wound to the contralateral arm was untreated and both wounds left to heal by secondary intention. Wounds were re-excised during the inflammatory phase (7 days, n = 10), matrix remodelling (3 weeks, n = 8) and cosmetic outcome/dermal structure (9 months, n = 9). Production of TGF-β1, TGF-β3 and matrix metalloproteinases (MMPs) was assessed by immunohistochemistry alongside microscopic measurement of wound size/area and clinical assessment of wound appearance. MAL-PDT delayed re-epithelialization at 7 days, associated with increased inflammation. However, 3 weeks postwounding, treated wounds were smaller with higher production of MMP-1 (P = 0·01), MMP-9 (P = 0·04) and TGF-β3 (P = 0·03). TGF-β1 was lower than control at 7 days and higher at 3 weeks (both P = 0·03). At 9 months, MAL-PDT-treated wounds showed greater, more ordered deposition of collagen I, collagen III and elastin (all P < 0·05). MAL-PDT increases MMP-1, MMP-9 and TGF-β3 production during matrix remodelling, ultimately producing scars with improved dermal matrix architecture.