EMAIL THIS PAGE TO A FRIEND
Genome announcements
Draft Genome Sequence of Sphingobium sp. Strain C100, a Polycyclic Aromatic Hydrocarbon-Degrading Bacterium from the Deep-Sea Sediment of the Arctic Ocean.
PMID 24482512
Abstract
Sphingobium sp. strain C100 was isolated from a polycyclic aromatic hydrocarbon (PAH)-degrading consortium from the deep-sea sediment of the Arctic Ocean. It can degrade two- to four-ring PAHs at 25°C. Here we present the draft genome sequence of this strain, which is 4,776,810xa0bp with a G+C content of 63.9%.