EMAIL THIS PAGE TO A FRIEND
Cold Spring Harbor perspectives in biology
Cellular reprogramming.
PMID 24492711
Abstract
Nuclear reprogramming technology was first established more than 50 years ago. It can rejuvenate somatic cells by erasing the epigenetic memories and reconstructing a new pluripotent order. The recent discovery reviewed here that induced pluripotency can be achieved by a small set of transcription factors has opened up unprecedented opportunities in the pharmaceutical industry, the clinic, and laboratories. This technology allows us to access pathological studies by using patient-specific induced pluripotent stem (iPS) cells. In addition, iPS cells are also expected to be a rising star for regenerative medicine, as sources of transplantation therapy.