EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Oclacitinib (APOQUEL(®)) is a novel Janus kinase inhibitor with activity against cytokines involved in allergy.
PMID 24495176
Abstract
Janus kinase (JAK) enzymes are involved in cell signaling pathways activated by various cytokines dysregulated in allergy. The objective of this study was to determine whether the novel JAK inhibitor oclacitinib could reduce the activity of cytokines implicated in canine allergic skin disease. Using isolated enzyme systems and in vitro human or canine cell models, potency and selectivity of oclacitinib was determined against JAK family members and cytokines that trigger JAK activation in cells. Oclacitinib inhibited JAK family members by 50% at concentrations (IC50 's) ranging from 10 to 99 nm and did not inhibit a panel of 38 non-JAK kinases (IC50 's > 1000 nM). Oclacitinib was most potent at inhibiting JAK1 (IC50 = 10 nM). Oclacitinib also inhibited the function of JAK1-dependent cytokines involved in allergy and inflammation (IL-2, IL-4, IL-6, and IL-13) as well as pruritus (IL-31) at IC50 's ranging from 36 to 249 nM. Oclacitinib had minimal effects on cytokines that did not activate the JAK1 enzyme in cells (erythropoietin, granulocyte/macrophage colony-stimulating factor, IL-12, IL-23; IC50 's > 1000 nM). These results demonstrate that oclacitinib is a targeted therapy that selectively inhibits JAK1-dependent cytokines involved in allergy, inflammation, and pruritus and suggests these are the mechanisms by which oclacitinib effectively controls clinical signs associated with allergic skin disease in dogs.