EMAIL THIS PAGE TO A FRIEND
Mutation research. Genetic toxicology and environmental mutagenesis
Cytotoxicity and genotoxicity of orthodontic bands with or without silver soldered joints.
PMID 24495929
Abstract
Stainless steel bands, with or without silver soldered joints, are routinely used in orthodontics. However, little is known about the toxic biological effects of these appliances. The aims of this study were to evaluate the cytotoxic, cytostatic, genotoxic and DNA damage-inducing effects of non-soldered bands (NSB) and silver soldered bands (SSB) on the HepG2 and HOK cell lines and to quantify the amount of ions released by the bands. The 24-h metallic eluates of NSBs and SSBs were quantified by atomic absorption spectrophotometry. An MTT reduction assay was performed to evaluate the cytotoxicity, alkaline and modified comet assays were employed to measure genotoxicity and oxidative DNA damage effects, and cytokinesis-block micronucleus cytome (CBMN-Cyt) assays were used to verify DNA damage, cytostasis and cytotoxicity. Ag, Cd, Cr, Cu and Zn were detected in SSB medium samples, and Fe and Ni were detected in both the SSB and NSB medium samples. The SSB group induced stronger cytotoxic effects than the NSB group in both evaluated cell lines. NSB and SSB induced genotoxicity as evaluated by comet assays; stronger effects were observed in the SSB group. Both groups induced similar increases in the number of oxidative DNA lesions, as detected by the FPG and Endo III enzymes. Nucleoplasmic bridges, biomarkers of DNA misrepair and/or telomere end fusions, were significantly elevated in the SSB group. The SSB eluates showed higher amounts of Ni and Fe than NSB, and all the quantified ions were detected in SSB eluates, including Cd. The SSB eluates were more cytotoxic and genotoxic than the NSB samples. Based on these results, we propose that other brands, materials and techniques should be further investigated for the future manufacture of orthodontic appliances.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7000
1-Naphthyl phosphate monosodium salt monohydrate, ≥98% (titration), powder
C10H8NaO4P · H2O
C9157
Activated charcoal, suitable for cell culture, suitable for plant cell culture
C
C6289
Activated charcoal, plant cell culture tested
C
390127
Activated charcoal, purified by resistive heating of graphite
C
05112
Activated charcoal, for GC
C
05105
Activated charcoal, powder
C
31616
Activated charcoal, puriss. p.a., powder
C
80485
Activated charcoal, for the determination of AOX, 50-150 μm particle size
C
440345
Glass spheres, 9-13 μm particle size
O2Si
420875
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
O2Si
420832
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420840
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420883
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
O2Si
420891
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
O2Si
420824
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420816
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420808
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
O2Si
420794
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
O2Si
420786
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420778
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
O2Si
420859
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
O2Si
274739
Sand, white quartz, 50-70 mesh particle size
O2Si
83340
Sand, white quartz, purum p.a., powder
O2Si
204358
Sand, white quartz, ≥99.995% trace metals basis
O2Si
421553
Silica
O2Si
381276
Silica, 99.8%
O2Si
718483
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
O2Si
748161
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
O2Si
S5505
Silica, fumed, powder, 0.2-0.3 μm avg. part. size (aggregate)
O2Si
S5130
Silica, fumed, powder
O2Si