EMAIL THIS PAGE TO A FRIEND
Inflammation
Rosiglitazone dampens pulmonary inflammation in a porcine model of acute lung injury.
PMID 24497162
Abstract
The hallmarks of acute lung injury (ALI) are the compromised alveolar-capillary barrier and the extravasation of leukocytes into the alveolar space. Given the fact that the peroxisome proliferator-activated receptor-γ agonist rosiglitazone holds significant anti-inflammatory properties, we aimed to evaluate whether rosiglitazone could dampen these hallmarks of local pulmonary inflammation in a porcine model of lung injury. For this purpose, we used a model of lipopolysaccharide (LPS, 50xa0μg/kg)-induced ALI. One hundred twenty minutes following the infusion of LPS, we started the exposure to rosiglitazone through inhalation or infusion. We found that intravenous rosiglitazone significantly controlled local pulmonary inflammation as determined through the expression of cytokines within the alveolar compartment. Furthermore, we found a significant reduction of the protein concentration and neutrophil activity within the alveolar space. In summary, we therefore conclude that the treatment with rosiglitazone might dampen local pulmonary inflammation during the initial stages of ALI.