EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Validation of genome-wide association study (GWAS)-identified disease risk alleles with patient-specific stem cell lines.
PMID 24497574
Abstract
While the past decade has seen great progress in mapping loci for common diseases, studying how these risk alleles lead to pathology remains a challenge. Age-related macular degeneration (AMD) affects 9 million older Americans, and is characterized by the loss of the retinal pigment epithelium (RPE). Although the closely linked genome-wide association studies ARMS2/HTRA1 genes, located at the chromosome 10q26 locus, are strongly associated with the risk of AMD, their downstream targets are unknown. Low population frequencies of risk alleles in tissue banks make it impractical to study their function in cells derived from autopsied tissue. Moreover, autopsy eyes from end-stage AMD patients, where age-related RPE atrophy and fibrosis are already present, cannot be used to determine how abnormal ARMS2/HTRA1 expression can initiate RPE pathology. Instead, induced pluripotent stem (iPS) cell-derived RPE from patients provides us with earlier stage AMD patient-specific cells and allows us to analyze the underlying mechanisms at this critical time point. An unbiased proteome screen of A2E-aged patient-specific iPS-derived RPE cell lines identified superoxide dismutase 2 (SOD2)-mediated antioxidative defense in the genetic allele's susceptibility of AMD. The AMD-associated risk haplotype (T-in/del-A) impairs the ability of the RPE to defend against aging-related oxidative stress. SOD2 defense is impaired in RPE homozygous for the risk haplotype (T-in/del-A; T-in/del-A), while the effect was less pronounced in RPE homozygous for the protective haplotype (G-Wt-G; G-Wt-G). ARMS2/HTRA1 risk alleles decrease SOD2 defense, making RPE more susceptible to oxidative damage and thereby contributing to AMD pathogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405779
Anti-F3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410355
Anti-F3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA001527
Anti-TF antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005692
Anti-TF antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW20009F
Anti-Transferrin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
T2027 Anti-Transferrin antibody produced in goat, whole antiserum
L7757
Lidocaine, powder
C14H22N2O
L1026
Lidocaine, analytical standard
C14H22N2O
PHR1034
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
C14H22N2O
L0595000
Lidocaine, European Pharmacopoeia (EP) Reference Standard
C14H22N2O
1366002
Lidocaine, United States Pharmacopeia (USP) Reference Standard
C14H22N2O
WH0002152M1
Monoclonal Anti-F3 antibody produced in mouse, clone 4G4, purified immunoglobulin, buffered aqueous solution
SAB1403798
Monoclonal Anti-F3 antibody produced in mouse, clone 4G4, ascites fluid
SAB1402940
Monoclonal Anti-TF antibody produced in mouse, clone 1C2, purified immunoglobulin, buffered aqueous solution