EMAIL THIS PAGE TO A FRIEND
PLoS genetics
WNT7B promotes bone formation in part through mTORC1.
PMID 24497849
Abstract
WNT signaling has been implicated in both embryonic and postnatal bone formation. However, the pertinent WNT ligands and their downstream signaling mechanisms are not well understood. To investigate the osteogenic capacity of WNT7B and WNT5A, both normally expressed in the developing bone, we engineered mouse strains to express either protein in a Cre-dependent manner. Targeted induction of WNT7B, but not WNT5A, in the osteoblast lineage dramatically enhanced bone mass due to increased osteoblast number and activity; this phenotype began in the late-stage embryo and intensified postnatally. Similarly, postnatal induction of WNT7B in Runx2-lineage cells greatly stimulated bone formation. WNT7B activated mTORC1 through PI3K-AKT signaling. Genetic disruption of mTORC1 signaling by deleting Raptor in the osteoblast lineage alleviated the WNT7B-induced high-bone-mass phenotype. Thus, WNT7B promotes bone formation in part through mTORC1 activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1300743 Anti-LRP6 (center) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1300317 Anti-LRP6 (N-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
L9294
Anti-LRP6 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA029925
Anti-LRP6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300444
Anti-RPS6KB1 (Ab-389) antibody produced in rabbit, affinity isolated antibody
SAB4300515
Anti-RPS6KB1 (Ab-411) antibody produced in rabbit, affinity isolated antibody
SAB4300500
Anti-RPS6KB1 (Ab-421) antibody produced in rabbit, affinity isolated antibody
SAB4300527
Anti-RPS6KB1 (Ab-424) antibody produced in rabbit, affinity isolated antibody
SAB1412617
ANTI-RPS6KB1 antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution
SAB1412626
ANTI-RPS6KB1 antibody produced in mouse, clone 4H4, purified immunoglobulin, buffered aqueous solution
SAB2104784
Anti-RPS6KB1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2104785
Anti-RPS6KB1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
S4047 Anti-S6 Kinase antibody produced in rabbit, whole antiserum
SAB4502691
Anti-S6K antibody produced in rabbit, affinity isolated antibody
02685
Lithium atomic spectroscopy standard concentrate 1.00 g Li, for 1 l standard solution, 1.00 g/L, analytical standard
ClLi
746460
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
ClLi
793620
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
ClLi
L4408
Lithium chloride, BioXtra, ≥99.0% (titration)
ClLi
62476
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
ClLi
L9650
Lithium chloride, for molecular biology, ≥99%
ClLi
429457
Lithium chloride, anhydrous, beads, −10 mesh, 99.998% trace metals basis
ClLi
449881
Lithium chloride, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
ClLi
203637
Lithium chloride, powder, ≥99.99% trace metals basis
ClLi
HT-LICLSLUG
Lithium chloride, anhydrous, chunks, 99.99% trace metals basis
ClLi
751898
Lithium chloride, anhydrous, chunks, 99.99% trace metals basis
ClLi
L7026
Lithium chloride solution, 8 M, for molecular biology, ≥99%
ClLi
694576
Lithium chloride solution, 1 M in 1,3-dimethyl-2-imidazolidinone
ClLi
62483
Lithium chloride solution, 1 M in glacial acetic acid
ClLi
62481
Lithium chloride solution, 1 M in ethanol
ClLi
667544
Lithium chloride solution, 0.5 M in anhydrous tetrahydrofuran
ClLi