EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
The known interactome of PTPIP51 in HaCaT cells—inhibition of kinases and receptors.
PMID 24501773
Abstract
Protein tyrosine phosphatse interacting protein (PTPIP51) is involved in the modulation of the mitogen activated protein kinase (MAPK) signaling pathway. Up to now, less is known about the regulation of this modulation. A recent study hinted to the phosphorylation status of PTPIP51 being essential for correct regulation of PTPIP51 function.In this study we investigate the phosphorylation status of PTPIP51 under the inhibition of the main interacting kinases and phosphatases of PTPIP51. c-Src was inhibited by Dasatinib, EGF receptor by Gefitinib, protein kinase C by staurosporine, protein kinase A by RpcAMPs and PTP1B by its specific inhibitor. Furthermore, a combination of PP2 with Gefitinib and RpcAMPs was used, respectively. The data were acquired for non-EGF and EGF treated HaCaT cells.All cells were analyzed relative to the subcellular distribution and change in the amount of tyrosine 176 phosphorylated PTPIP51. Furthermore, the protein interactions were assayed by duolink proximity ligation assay.HaCaT cells submitted to the respective inhibitor displayed a subcellular redistribution of tyrosine 176 phosphorylated PTPIP51 depending on the applied inhibitor. Yet, the amount of tyrosine 176 phosphorylated PTPIP51 remained unchanged by inhibitor treatment except for Gefitinib and simultaneous PP2 and Gefitnib treatment in non EGF-stimulated cells, but was elevated if cells were also EGF stimulated, in control and inhibitor treated cells. Interestingly, the interaction with EGFR, 14-3-3, Raf-1, c-Src, PTP1B, PKA and PKC was influenced by the application of inhibitors. Also EGF application resulted in a sharp drop of the PTPIP51 interaction with the MAPK pathway (e.g. Raf-1) in the control group.Summarizing these new findings, we postulate that PTPIP51 is regulated by its phosphorylation status combined with a thereby induced subcellular redistribution. In addition, the EGF receptor regulates PTPIP51 interaction with Raf-1 by its phosphorylation, thus preventing an overshooting activation of the MAPK pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA009975
Anti-RMDN3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
DUO92012
Duolink® In Situ Detection Reagents Brightfield
DUO92013
Duolink® In Situ Detection Reagents FarRed
DUO92014
Duolink® In Situ Detection Reagents Green
DUO92007
Duolink® In Situ Detection Reagents Orange
DUO92008
Duolink® In Situ Detection Reagents Red
DUO92106
Duolink® In Situ Orange Starter Kit Goat/Rabbit
DUO92104
Duolink® In Situ Orange Starter Kit Mouse/Goat
DUO92102
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
DUO92006
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92003
Duolink® In Situ PLA® Probe Anti-Goat PLUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92004
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92001
Duolink® In Situ PLA® Probe Anti-Mouse PLUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92005
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92002
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92010 Duolink® In Situ Probemaker MINUS
DUO92009 Duolink® In Situ Probemaker PLUS
DUO92105
Duolink® In Situ Red Starter Kit Goat/Rabbit
DUO92103
Duolink® In Situ Red Starter Kit Mouse/Goat
DUO92101
Duolink® In Situ Red Starter Kit Mouse/Rabbit