EMAIL THIS PAGE TO A FRIEND
Arthritis & rheumatology (Hoboken, N.J.)
Brief report: Autologous stem cell transplantation restores immune tolerance in experimental arthritis by renewal and modulation of the Teff cell compartment.
PMID 24504807
Abstract
Autologous stem cell transplantation (ASCT) induces long-term drug-free disease remission in patients with juvenile idiopathic arthritis. This study was undertaken to further unravel the immunologic mechanisms underlying ASCT by using a mouse model of proteoglycan-induced arthritis (PGIA). For initiation of PGIA, BALB/c mice received 2 intraperitoneal injections of human PG in a synthetic adjuvant on days 0 and 21. Five weeks after the first immunization, the mice were exposed to total body irradiation (7.5 Gy) and received (un)manipulated bone marrow (BM) grafts from mice with PGIA. Clinical scores, T cell reconstitution, (antigen-specific) T cell cytokine production, and intracellular cytokine expression were determined following autologous BM transplantation (ABMT). ABMT resulted in amelioration and stabilization of arthritis scores. BM grafts containing T cells and T cell-depleted grafts provided the same clinical benefit, with similar reductions in PG-induced T cell proliferation and the number of PG-specific autoantibodies. In vivo reexposure to PG did not exacerbate disease. Following ABMT, basal levels of disease-associated proinflammatory cytokines (interferon-γ [IFNγ], interleukin-17 [IL-17], and tumor necrosis factor α [TNFα]) were reduced. In addition, restimulation of T cells with PG induced a strong reduction in disease-associated proinflammatory cytokine production. Finally, although the remaining host T cells displayed a proinflammatory phenotype following ABMT, IFNγ, IL-17, and TNFα production by the newly reconstituted donor-derived T cells was significantly lower. Taken together, our data suggest that ABMT restores immune tolerance by renewal and modulation of the Teff cell compartment, leading to a strong reduction in proinflammatory (self antigen-specific) T cell cytokine production.