PloS one
Paradoxical roles of mineral dust induced gene on cell proliferation and migration/invasion.
PMID 24505346
Abstract
Increased expression of mineral dust-induced gene (mdig, also named as mina53, MINA, or NO52) has been observed in a number of human cancers. The mechanism of how mdig contribute to the pathogenesis of cancer remains to be fully elucidated. In this report, we demonstrated that overexpression of mdig decreased the nuclear staining signal by 4',6-diamidino-2-phenylindole (DAPI), along with a considerable enhancement in cell proliferation. Silencing mdig by shRNA resulted in a statistically significant decrease of cell proliferation. Intriguingly, mdig overexpression reduced the capacity of the cells in migration and invasion in vitro, whereas silencing mdig by shRNA/siRNA enhanced migration and invasion. Clinically, we found that increased expression of mdig in cancer tissues correlates with poorer overall survival of the lung cancer patients, esp., for those without lymph node metastasis. Taken together, our results suggest that mdig plays opposite roles on cell growth and motility, which possibly indicates the paradoxical effect of mdig at the different stages of carcinogenesis.