EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Alzheimer's disease therapeutics targeted to the control of amyloid precursor protein translation: maintenance of brain iron homeostasis.
PMID 24513321
Abstract
The neurotoxicity of amyloid beta (Aβ), a major cleavage product of the amyloid precursor protein (APP), is enhanced by iron, as found in the amyloid plaques of Alzheimer's disease (AD) patients. By contrast, the long-known neuroprotective activity of APP is evident after α-secretase cleavage of the precursor to release sAPPα, and depends on the iron export actions of APP itself. The latter underlie its neurotrophic and protective effects in facilitating the homeostatic actions of ferroportin mediated-iron export. Thus APP-dependent iron export may alleviate oxidative stress by minimizing labile iron thus protecting neurons from iron overload during stroke and hemorrhage. Consistent with this, altered phosphorylation of iron-regulatory protein-1 (IRP1) and its signaling processes play a critical role in modulating APP translation via the 5' untranslated region (5'UTR) of its transcript. The APP 5'UTR region encodes a functional iron-responsive element (IRE) RNA stem loop that represents a potential target for modulating APP production. Targeted regulation of APP gene expression via the modulation of 5'UTR sequence function represents a novel approach for the potential treatment of AD since altering APP translation can be used to improve both the protective brain iron balance and provide anti-amyloid efficacy. Approved drugs including paroxetine and desferrioxamine and several novel compounds have been identified that suppress abnormal metal-promoted Aβ accumulation with a subset of these acting via APP 5'UTR-dependent mechanisms to modulate APP translation and cleavage to generate the non-toxic sAPPα.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF58945078
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%
Fe
GF21695722
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Fe
GF81282135
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Fe
GF57565996
Iron, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Fe
GF48022152
Iron, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Fe
GF48020947
Iron, wire reel, 500m, diameter 0.125mm, hard, 99.5%
Fe
GF58809965
Iron, wire reel, 50m, diameter 0.065mm, hard, 99.5%
Fe
GF34071566
Iron, wire reel, 50m, diameter 0.25mm, hard, 99.5%
Fe
GF96915134
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Fe
GF35663935
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Fe
GF27511781
Iron, wire reel, 5m, diameter 0.25mm, annealed, 99.99+%
Fe
GF61040023
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Fe
GF55306964
Iron, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Fe
GF92087242
Iron, wire reel, 5m, diameter 0.5mm, as drawn, 99.95%
Fe
GF93388999
Iron, foil, light tested, 25x25mm, thickness 0.075mm, hard, 99.5%
Fe
GF90779899
Iron, foil, light tested, 50x50mm, thickness 0.01mm, 99.99+%
Fe
GF09386084
Iron, foil, light tested, 50x50mm, thickness 0.075mm, as rolled, 99.99+%
Fe
GF18004115
Iron, foil, not light tested, 100x100mm, thickness 0.005mm, 99.85%
Fe
GF30775248
Iron, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.99+%
Fe
GF25243174
Iron, foil, not light tested, 150x150mm, thickness 0.003mm, 99.85%
Fe
GF83094824
Iron, rod, 300mm, diameter 6.0mm, as drawn, 99.99+%
Fe
GF97671415
Iron, rod, 500mm, diameter 4.8mm, as drawn, 98+%
Fe
GF19311724
Iron, rod, 500mm, diameter 9.5mm, as drawn, 98+%
Fe
GF91822653
Iron, rod, 50mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Fe
GF39505518
Iron, tube, 1000mm, outside diameter 1.40mm, inside diameter 0.8mm, wall thickness 0.3mm, as drawn, 99.8%
Fe
GF62180928
Iron, wire reel, 2000m, diameter 0.065mm, hard, 99.5%
Fe
GF05699975
Iron, wire reel, 250m, diameter 0.065mm, hard, 99.5%
Fe
GF56751206
Iron, wire reel, 2m, diameter 1.0mm, as drawn, 99.99+%
Fe
GF01815702
Iron, wire reel, 5m, diameter 0.13mm, hard, 99.99+%
Fe
GF69332519
Iron, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Fe