EMAIL THIS PAGE TO A FRIEND
Advanced healthcare materials
Lung deposition and cellular uptake behavior of pathogen-mimicking nanovaccines in the first 48 hours.
PMID 24520022
Abstract
Pulmonary immunization poses the unique challenge of balancing vaccine efficacy with minimizing inflammation in the respiratory tract. While previous studies have shown that mice immunized intranasally with F1-V-loaded polyanhydride nanoparticles are protected from a lethal challenge with Yersinia pestis, little is known about the initial interaction between the nanoparticles and immune cells following intranasal administration. Here, the deposition within the lung and internalization by phagocytic cells of polyanhydride nanovaccines encapsulating F1-V are compared with that of soluble F1-V alone or F1-V adjuvanted with monophosphoryl lipid A (MPLA). Encapsulation of F1-V into polyanhydride nanoparticles prolonged its presence while F1-V administered with MPLA is undetectable within 48 h. The inflammation induced by the polyanhydride nanovaccine is mild compared with the marked inflammation induced by the MPLA-adjuvanted F1-V. Even though F1-V delivered with saline is detected in the lung 48 h after administration, it is known that this regimen does not elicit a protective immune response. The prolonged F1-V presence in the lung in concert with the mild inflammatory response provided by the nanovaccine provides new insights into the development of protective immune responses with a single intranasal dose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
691275
Acetic anhydride, Lonza quality, ≥99.5% (GC)
C4H6O3
242845
Acetic anhydride, ACS reagent, ≥98.0%
C4H6O3
320102
Acetic anhydride, ReagentPlus®, ≥99%
C4H6O3
539996
Acetic anhydride, 99.5%
C4H6O3
A6404
Acetic anhydride, reagent grade, ≥98%
C4H6O3
45840
Acetic anhydride
C4H6O3
01-0390
Acetic anhydride, SAJ first grade, ≥93.0%
C4H6O3
01-0380
Acetic anhydride, JIS special grade
C4H6O3
L095004
Acetic anhydride
C4H6O3
91238
Diethyl ether, analytical standard
C4H10O
09-1833
Diethyl ether, for residue analysis, suitable for 5000 per JIS
C4H10O
09-1855
Diethyl ether
C4H10O
236705
Pentane, anhydrous, ≥99%
C5H12
34956
Pentane, for HPLC, ≥99.0%
C5H12
154954
Pentane, spectrophotometric grade, ≥99%
C5H12
158941
Pentane, reagent grade, 98%
C5H12
76870
Pentane, analytical standard
C5H12
76871
Pentane, puriss. p.a., ≥99.0% (GC)
C5H12
60489
Pentane, ≥99% (GC)
C5H12
76874
Pentane, purum, ≥95.0% (GC)
C5H12
442746
Pentane, analytical standard
C5H12
791776
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
CK2O3
P5833
Potassium carbonate, BioXtra, ≥99.0%
CK2O3
P1472
Potassium carbonate, meets USP testing specifications
CK2O3
367877
Potassium carbonate, 99.995% trace metals basis
CK2O3
590681
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
CK2O3
209619
Potassium carbonate, ACS reagent, ≥99.0%
CK2O3
310263
Potassium carbonate, ReagentPlus®, 99%
CK2O3
347825
Potassium carbonate, reagent grade, ≥98%, powder, −325 mesh
CK2O3
60108
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
CK2O3