EMAIL THIS PAGE TO A FRIEND
Pharmaceutical research
Novel nanostructured lipid carrier co-loaded with doxorubicin and docosahexaenoic acid demonstrates enhanced in vitro activity and overcomes drug resistance in MCF-7/Adr cells.
PMID 24522814
Abstract
To develop a nanostructured lipid carrier (NLC) co-loaded with doxorubicin and docosahexaenoic acid (DHA) and to evaluate its potential to overcome drug resistance and to increase antitumoral effect in MCF-7/Adr cancer cell line. The NLC was prepared by a hot homogenization method and characterized for size, zeta potential, entrapment efficiency (EE) and drug loading (DL). Drug release was evaluated by dialysis in complete DMEM, and NLC aggregation was assayed in the presence of serum. The cytotoxicity of formulations, doxorubicin uptake or penetration were evaluated in MCF-7 and MCF-7/Adr as monolayer or spheroid models. The formulation had a size of about 80xa0nm, negative zeta potential, EE of 99%, DL of 31xa0mg/g, a controlled drug release in DMEM and no particles aggregation in presence of serum. The NLC loaded with doxorubicin and DHA showed the same activity as free drugs against MCF-7 but a stronger activity against MCF-7/Adr cells. In monolayer model, the doxorubicin uptake as free and encapsulated form was similar in MCF-7 but higher for the encapsulated drug in MCF-7/Adr, suggesting a bypassing of P-glycoprotein bomb efflux. For spheroids, the NLC loaded with doxorubicin and DHA showed a prominent cytotoxicity and a greater penetration of doxorubicin. These findings suggest that the co-encapsulation of doxorubicin and DHA in NLC enhances the cytotoxicity and overcomes the doxorubicin resistance in MCF-7/Adr.