EMAIL THIS PAGE TO A FRIEND
British journal of clinical pharmacology
Pharmacodynamics and pharmacokinetics during the transition from warfarin to rivaroxaban: a randomized study in healthy subjects.
PMID 24528331
Abstract
This study investigated relevant pharmacodynamic and pharmacokinetic parameters during the transition from warfarin to rivaroxaban in healthy male subjects. Ninety-six healthy men were randomized into the following three groups: warfarin [international normalized ratio (INR) 2.0-3.0] transitioned to rivaroxaban 20 mg once daily (od; group A); warfarin (INR 2.0-3.0) followed by placebo od (group B); and rivaroxaban alone 20 mg od (group C) for 4 days. Anti-factor Xa activity, inhibition of factor Xa activity, prothrombin time (PT), activated partial thromboplastin time, HepTest, prothrombinase-induced clotting time, factor VIIa activity, factor IIa activity, endogenous thrombin potential and pharmacokinetics were measured. An additive effect was observed on the PT and PT/INR during the initial transition period. The mean maximal prolongation of PT was 4.39-fold [coefficient of variation (CV) 18.03%; range 3.39-6.50] of the baseline value in group A, compared with 1.88-fold (CV 10.35%; range 1.53-2.21) in group B and 1.57-fold (CV 9.98%; range 1.37-2.09) in group C. Rivaroxaban had minimal influence on the PT/INR at trough levels. Inhibition of factor Xa activity, activated partial thromboplastin time and endogenous thrombin potential were also enhanced, but to a lesser extent. In contrast, the effects of rivaroxaban on anti-factor Xa activity, HepTest and prothrombinase-induced clotting time were not affected by pretreatment with warfarin. Changes in pharmacodynamics during the transition from warfarin to rivaroxaban vary depending on the test used. A supra-additive effect on PT/INR is expected during the initial period of transition, but pretreatment with warfarin does not influence the effect of rivaroxaban on anti-factor Xa activity.