EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Aging-related dysregulation of dopamine and angiotensin receptor interaction.
PMID 24529758
Abstract
It is not known whether the aging-related decrease in dopaminergic function leads to the aging-related higher vulnerability of dopaminergic neurons and risk for Parkinson's disease. The renin-angiotensin system (RAS) plays a major role in the inflammatory response, neuronal oxidative stress, and dopaminergic vulnerability via type 1 (AT1) receptors. In the present study, we observed a counterregulatory interaction between dopamine and angiotensin receptors. We observed overexpression of AT1 receptors in the striatum and substantia nigra of young adult dopamine D1 and D2 receptor-deficient mice and young dopamine-depleted rats, together with compensatory overexpression of AT2 receptors or compensatory downregulation of angiotensinogen and/or angiotensin. In aged rats, we observed downregulation of dopamine and dopamine receptors and overexpression of AT1 receptors in aged rats, without compensatory changes observed in young animals. L-Dopa therapy inhibited RAS overactivity in young dopamine-depleted rats, but was ineffective in aged rats. The results suggest that dopamine may play an important role in modulating oxidative stress and inflammation in the substantia nigra and striatum via the RAS, which is impaired by aging.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H8523
6-Hydroxydopamine hydrobromide, ≥90% (HPLC), solid
C8H11NO3 · HBr
H116
6-Hydroxydopamine hydrobromide, contains ascorbic acid as stabilizer, ≥98% (HPLC)
C8H11NO3 · HBr
162957
6-Hydroxydopamine hydrobromide, 95%
C8H11NO3 · HBr
GW22763 Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500247
Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405848
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701826
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701825
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103104
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4300645
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody
HPA040067
Anti-GAPDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PLA0302
Goat anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
WH0002597M1
Monoclonal Anti-GAPDH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
PLA0125
Rabbit anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.