EMAIL THIS PAGE TO A FRIEND
Reproduction (Cambridge, England)
Maternal thyroid dysfunction affects placental profile of inflammatory mediators and the intrauterine trophoblast migration kinetics.
PMID 24534949
Abstract
The objective of the present study was to evaluate the gene and immunohistochemical expression of inflammatory mediators involved in the immune activity and the intrauterine trophoblast migration of the placentas in hypothyroid and L-thyroxine (L-T4)-treated rats. A total of 144 adult female rats were divided equally into hypothyroid, l-T4-treated, and euthyroid (control) groups. Hypothyroidism was induced by daily administration of propylthiouracil. Rats were killed at 0, 10, 14, 15, 16, 17, 18, and 19 days of gestation. We evaluated the depth of interstitial and endovascular intrauterine trophoblast invasion and the immunohistochemical expression of interferon γ (INFy), migration inhibitory factor (MIF), and inducible nitric oxide synthase (NOS2 (iNOS)). The gene expression of Toll-like receptor 2 (Tlr2) and Tlr4, Infy, Mif, tumor necrosis factor (Tnf (Tnfα)), Il10, Nos2, matrix metalloproteinase 2 (Mmp2) and Mmp9, and placental leptin was also measured in placental disks by real-time RT-PCR. The data were analyzed using an Student-Newman-Keuls (SNK) test. Hypothyroidism reduced the endovascular and interstitial trophoblast migration, and the expression of TLR4, INFy, MIF, interleukin 10 (IL10), NOS2, MMP2 and MMP9, and placental leptin, while increased the expression of TLR2 (P<0.05). T4-treated rats not only increased the expression of IL10 and NOS2 but also reduced the expression of TNF and MIF at 10 days of gestation (P<0.05). However, at 19 days of gestation, expression of INFy and MIF was increased in T4-treated group (P<0.05). Excess of T4 also increased the gene expression of Mmp2 at 10 days of gestation (P<0.05), but reduced the endovascular trophoblast migration at 18 days of gestation (P<0.05). Hypothyroidism and excess of T4 differentially affect the immune profile and the intrauterine trophoblast migration of the placenta, and these effects are dependent on the gestational period.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405869
Anti-GIF antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA039908
Anti-GIF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA040774
Anti-GIF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410525
Anti-GIF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2500635
Anti-MIF antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA003868
Anti-MIF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410786
Anti-MIF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
N7782
Anti-Nitric Oxide Synthase, Inducible antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306705
ANTI-NOS2A (CENTER) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305735
ANTI-NOS2A(N-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
WH0003426M1
Monoclonal Anti-IF antibody produced in mouse, clone 1B3, purified immunoglobulin, buffered aqueous solution
WH0004282M1
Monoclonal Anti-MIF antibody produced in mouse, clone 2A10-4D3, purified immunoglobulin, buffered aqueous solution
N9657
Monoclonal Anti-Nitric Oxide Synthase, Inducible antibody produced in mouse, clone NOS-IN, ascites fluid
SAB1402859
Monoclonal Anti-NOS2 antibody produced in mouse, clone 2G4, purified immunoglobulin, buffered aqueous solution