EMAIL THIS PAGE TO A FRIEND
European journal of immunology
Lnk prevents inflammatory CD8⁺ T-cell proliferation and contributes to intestinal homeostasis.
PMID 24536025
Abstract
The intracellular adaptor Lnk (also known as SH2B3) regulates cytokine signals that control lymphohematopoiesis, and Lnk(-/-) mice have expanded B-cell, megakaryocyte, and hematopoietic stem-cell populations. Moreover, mutations in the LNK gene are found in patients with myeloproliferative disease, whereas LNK polymorphisms have recently been associated with inflammatory and autoimmune diseases, including celiac disease. Here, we describe a previously unrecognized function of Lnk in the control of inflammatory CD8(+) T-cell proliferation and in intestinal homeostasis. Mature T cells from newly generated Lnk-Venus reporter mice had low but substantial expression of Lnk, whereas Lnk expression was downregulated during homeostatic T-cell proliferation under lymphopenic conditions. The numbers of CD44(hi) IFN-γ(+) CD8(+) effector or memory T cells were found to be increased in Lnk(-/-) mice, which also exhibited shortening of villi in the small intestine. Lnk(-/-) CD8(+) T cells survived longer in response to stimulation with IL-15 and proliferated even in nonlymphopenic hosts. Transfer of Lnk(-/-) CD8(+) T cells together with WT CD4(+) T cells into Rag2-deficient mice recapitulated a sign of villous abnormality. Our results reveal a link between Lnk and immune cell-mediated intestinal tissue destruction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4503853
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
SAB4504016
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4504015
Anti-phospho-Akt (pSer246) antibody produced in rabbit, affinity isolated antibody
SAB4504331
Anti-phospho-Akt (pSer473) antibody produced in rabbit, affinity isolated antibody
SAB4504332
Anti-phospho-Akt (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504017
Anti-phospho-Akt (pThr72) antibody produced in rabbit, affinity isolated antibody
SAB4504110
Anti-phospho-Akt (pTyr326) antibody produced in rabbit, affinity isolated antibody
SAB4504014
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody
SAB4300042
Anti-phospho-AKT1 (pSer473) antibody produced in rabbit, affinity isolated antibody
SAB4300043
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504749
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4503852
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody
SAB4300260
Anti-phospho-AKT1 (pThr450) antibody produced in rabbit, affinity isolated antibody
P3862 Anti-phospho-PKB (pThr308) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4300034
Anti-phospho-STAT3 (pSer727) antibody produced in rabbit, affinity isolated antibody
SAB4504540
Anti-phospho-STAT3 (pSer727) antibody produced in rabbit, affinity isolated antibody
S2690 Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300033
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
SAB4504541
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
HPA005483
Anti-SH2B3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution