EMAIL THIS PAGE TO A FRIEND
Journal of chemotherapy (Florence, Italy)
Comparative efficacy of tigecycline VERSUS vancomycin in an experimental model of soft tissue infection by methicillin-resistant Staphylococcus aureus producing Panton-Valentine leukocidin.
PMID 24548093
Abstract
Methicillin-resistant Staphylococcus aureus (MRSA) producing Panton-Valentine leukocidin (PVL) is highly virulent. This study aimed to compare the efficacy of tigecycline versus vancomycin in experimental thigh abscess by a PVL-producing MRSA isolate. One hundred and ninety-six Wistar rats were divided into five groups: group A, controls; groups B and C, administered vancomycin starting 1 and 6 h after bacterial challenge respectively; groups D and E, administered tigecycline starting 1 and 6 h after bacterial challenge respectively. Treatment was continued every 12 hours for three consecutive days. Survival was recorded; separate animals were killed for quantitative cultures. Serum samples were collected for estimation of malondialdehyde (MDA). Survival of group D was prolonged compared to all other groups. The bacterial load of blood, liver, spleen and lung was significantly decreased within group D compared to group B at 36 hours. Treatment with tigecycline was accompanied by significant reduction of serum MDA at 24 hours. Tigecycline is comparable to vancomycin for the treatment of soft tissue infections by PVL-producing MRSA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108383
1,1,3,3-Tetramethoxypropane, 99%
C7H16O4