EMAIL THIS PAGE TO A FRIEND
Cancer letters
Gold nanoparticles in breast cancer treatment: promise and potential pitfalls.
PMID 24556077
Abstract
Despite remarkable achievements in the treatment of breast cancer, some obstacles still remain. Gold nanoparticles may prove valuable in addressing these problems owing to their unique characteristics, including their enhanced permeability and retention in tumor tissue, their light absorbance and surface plasmon resonance in near-infrared light, their interaction with radiation to generate secondary electrons, and their ability to be conjugated with drugs or other agents. Herein, we discuss some basic concepts of gold nanoparticles, and early results from studies regarding their use in breast cancer, including toxicity and side effects. We also discuss these particles' potential clinical applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF43139538
Gold, insulated wire, 5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Au
GF92307922
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Au
GF11444611
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%
Au
GF80583701
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.009mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Au
GF06766713
Gold, insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%
Au
GF25294620
Gold, microfoil, disks, 10mm, thinness 0.01μm, specific density 20.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Au
GF46990974
Gold, microfoil, disks, 10mm, thinness 0.05μm, specific density 101.3μg/cm2, permanent mylar 3.5μm support, 99.99+%
Au
GF23782350
Gold, microfoil, disks, 10mm, thinness 0.1μm, specific density 204.1μg/cm2, permanent mylar 3.5μm support, 99.99+%
Au
GF38598980
Gold, microfoil, disks, 10mm, thinness 0.5μm, specific density 966μg/cm2, permanent mylar 3.5μm support, 99.99+%
Au
GF87786040
Gold, microfoil, disks, 10mm, thinness 0.75μm, specific density 1449μg/cm2, permanent mylar 3.5μm support, 99.99+%
Au
GF19229115
Gold, microfoil, disks, 10mm, thinness 1.0μm, specific density 1932μg/cm2, permanent mylar 3.5μm support, 99.99+%
Au
GF12231223
Gold, microleaf disks, 10mm, thinness 0.25μm, specific density 483μg/cm2, removable support, 99.99+%
Au
GF47078129
Gold, microleaf disks, 10mm, thinness 0.5μm, specific density 966μg/cm2, removable support, 99.99+%
Au
GF30409253
Gold, rod, 100mm, diameter 1.6mm, as drawn, 99.95%
Au
GF39029968
Gold, rod, 100mm, diameter 2.0mm, as drawn, 99.95%
Au
GF08843867
Gold, rod, 10mm, diameter 3.0mm, as drawn, 99.95%
Au
GF79016294
Gold, rod, 10mm, diameter 4.0mm, as drawn, 99.95%
Au
GF63042870
Gold, rod, 10mm, diameter 5.0mm, as drawn, 99.95%
Au
GF98454864
Gold, rod, 25mm, diameter 1.6mm, as drawn, 99.95%
Au
GF31080899
Gold, rod, 25mm, diameter 2.0mm, as drawn, 99.95%
Au
GF43173930
Gold, rod, 25mm, diameter 3.0mm, as drawn, 99.95%
Au
GF22371319
Gold, rod, 25mm, diameter 4.0mm, as drawn, 99.95%
Au
GF36535563
Gold, rod, 25mm, diameter 5.0mm, as drawn, 99.95%
Au
GF37867887
Gold, rod, 50mm, diameter 1.6mm, as drawn, 99.95%
Au
GF96742479
Gold, rod, 50mm, diameter 3.0mm, as drawn, 99.95%
Au
GF98812097
Gold, rod, 6mm, diameter 6.0mm, as drawn, 99.95%
Au
GF71552347
Gold, sphere, 100pcs, diameter 0.5mm, 99.99%
Au
GF80082479
Gold, sphere, 200pcs, diameter 0.5mm, 99.99%
Au
GF77657574
Gold, sphere, 20pcs, diameter 0.5mm, 99.99%
Au
GF73271280
Gold, tube, 100mm, outside diameter 0.3mm, inside diameter 0.2mm, wall thickness 0.05mm, as drawn, 99.95%
Au