EMAIL THIS PAGE TO A FRIEND
Vitamins and hormones
Roles of HNF1α and HNF4α in pancreatic β-cells: lessons from a monogenic form of diabetes (MODY).
PMID 24559927
Abstract
Mutations in the genes encoding hepatocyte nuclear factor (HNF)1α and HNF4α cause a monogenic form of diabetes mellitus known as maturity-onset diabetes of the young (MODY). The primary cause of MODY is an impairment of glucose-stimulated insulin secretion by pancreatic β-cells, indicating the important roles of HNF1α and HNF4α in β-cells. Large-scale genetic studies have clarified that the common variants of HNF1α and HNF4α genes are also associated with type 2 diabetes, suggesting that they are involved in the pathogenesis of both diseases. Recent experimental studies revealed that HNF1α controls both β-cell function and growth by regulating target genes such as glucose transporter 2, pyruvate kinase, collectrin, hepatocyte growth factor activator, and HNF4α. In contrast, HNF4α mainly regulates the function of β-cells. Although direct target genes of HNF4α in β-cells are largely unknown, we recently identified Anks4b as a novel target of HNF4α that regulates β-cell susceptibility to endoplasmic reticulum stress. Studies of MODY have led to a better understanding of the molecular mechanism of glucose-stimulated insulin secretion by pancreatic β-cells.