EMAIL THIS PAGE TO A FRIEND
Physiological research
Determinants of adrenal androgen hypofunction in premenopausal females with rheumatoid arthritis.
PMID 24564598
Abstract
The aim of our study was to investigate adrenocortical function in the context of disease activity and inflammatory status in premenopausal RA females. Adrenal glucocorticoid and androgen responses to the 1 microg ACTH 1-24 test were investigated in 23 premenopausal RA and in 15 age- and BMI-matched healthy females. Twelve RA patients were on low-dose prednisone (<8.5 mg/day). Patients with DAS28>3.2 had lower (p<0.05) total plasma cortisol, 17-hydroxyprogesterone, dehydroepiandrosterone and androstenedione responses in the ACTH test compared to healthy controls. Patients with DAS28>3.2 had lower (p<0.05) dehydroepiandrosterone response in the ACTH test compared to patients with DAS28