EMAIL THIS PAGE TO A FRIEND
Neuroscience research
Effect of phosphomimetic mutations on the C-terminal sensitivity of glycine transporter GlyT1 to calpain.
PMID 24566201
Abstract
Previously, we found that the C-terminus of the glycine transporter GlyT1 loses the most of its epitopes during pathological calcium increase in rodent synaptosomes but that the more internal epitopes are spared. We also found that epitope immunoreactivity likely decreases via both phosphorylation and calpain-mediated proteolysis. Here we show that the predicted phosphomimetic mutation S605D fully blocks the adjacent (T602/T603) internal calpain cleavage in the mouse GlyT1b C-terminal fusion protein, but that the neutral S605A mutation does not. Consistent with this, the phophomimetic mutation, but not the neutral mutation, significantly protected the internal GlyT1 C-terminal antiGlyT1C603-626 epitopes against calpain when introduced into tissue culture expressed GlyT1b. Because similar effects can be obtained using phosphatase inhibitors, it may be that phosphorylation of S605 protects the GlyT1 C-terminal sequences from calpain cleavage in vivo.