EMAIL THIS PAGE TO A FRIEND
Drug testing and analysis
Characterizing metabolites and potential metabolic pathways for the novel psychoactive substance methoxetamine.
PMID 24574323
Abstract
The classic approach of controlled volunteer studies to study drug metabolism is difficult or impossible to undertake for novel psychoactive substances (NPS), as there is generally very limited information available to allow appropriate dose finding and safety. A viable and powerful alternative is the identification and characterization of phase I and II metabolites of such drugs by examining the concordance of data gathered from analysis of microsomal incubates with that from analysis of specimens collected from individuals with analytically confirmed use of NPS. Liquid chromatography-high resolution mass spectrometry provides the ability to reliably identify such metabolites. We used this technique here to study the metabolism of the ketamine analogue methoxetamine. A large number of metabolites were identified in the in vitro studies including normethoxetamine, O-desmethylmethoxetamine, dihydromethoxetamine, dehydromethoxetamine and several structural isomers of hydroxymethoxetamine and hydroxynormethoxetamine. pH dependent liquid-liquid extraction was used to discriminate phenolic from alcoholic metabolites. Phase II glucuronide conjugates included those of O-desmethylmethoxetamine, O-desmethylnormethoxetamine and O-desmethylhydroxymethoxetamine. The majority of these phase I and II metabolites were confirmed to be present in urine collected from three individuals presenting with acute methoxetamine toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
414255
1-Chlorobutane, anhydrous, 99.5%
C4H9Cl
34958
1-Chlorobutane, for HPLC, ≥99.8%
C4H9Cl
09651
1-Chlorobutane, biotech. grade, for protein sequence analysis, ≥99.8%
C4H9Cl
125008
1-Chlorobutane, ReagentPlus®, 99%
C4H9Cl
71643
Sodium phosphate dibasic dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
HNa2O4P · 2H2O
30435
Sodium phosphate dibasic dihydrate, puriss. p.a., Reag. Ph. Eur., 98.5-101.0% (calc. to the dried substance)
HNa2O4P · 2H2O
04272
Sodium phosphate dibasic dihydrate, meets analytical specification of Ph. Eur., BP, 98.5-101% (calc. to the dried substance)
HNa2O4P · 2H2O
71645
Sodium phosphate dibasic dihydrate, purum p.a., ≥98.0% (T)
HNa2O4P · 2H2O
71662
Sodium phosphate dibasic dihydrate, tested according to Ph.Eur.
HNa2O4P · 2H2O
RES0974S-A7
Sodium phosphate dibasic dihydrate, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
HNa2O4P · 2H2O
71505
Sodium phosphate monobasic dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
H2NaO4P · 2H2O
71502
Sodium phosphate monobasic dihydrate, BioUltra, ≥99.0% (T)
H2NaO4P · 2H2O
71500
Sodium phosphate monobasic dihydrate, purum p.a., crystallized, ≥99.0% (T)
H2NaO4P · 2H2O
04269
Sodium phosphate monobasic dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, 98-100.5% (calc. to the dried substance)
H2NaO4P · 2H2O