EMAIL THIS PAGE TO A FRIEND
Medical mycology
Experimental murine acremoniosis: an emerging opportunistic human infection.
PMID 24577339
Abstract
Acremonium is an emerging fungal pathogen causing severe infections. We evaluated the virulence of three clinically relevant species within the genus, i.e., Acremonium kiliense (currently Sarocladium kiliense), Acremonium sclerotigenum-A. egyptiacum complex and Acremonium implicatum in a murine model of disseminated infection. Both immunocompetent and immunosuppresssed mice were infected with two inocula concentrations (2 × 10(6) and 2 × 10(8) conidia/animal) of two strains of each species. Tissue burden, mortality rate, histopathology and levels of (1→3)-β-D-glucan were used as virulence markers. None of the species of Acremonium tested was able to cause infection in immunocompetent mice. Conversely, severe infections were produced in immunocompromised mice, the spleen being the most affected organ. In general, the virulence of the Acremonium species tested was low, S. kiliense being the most virulent species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001046 Sevoflurane, European Pharmacopoeia (EP) Reference Standard
C4H3F7O