EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Downregulation of the tumor suppressor HSPB7, involved in the p53 pathway, in renal cell carcinoma by hypermethylation.
PMID 24585183
Abstract
In order to identify genes involved in renal carcinogenesis, we analyzed the expression profile of renal cell carcinomas (RCCs) using microarrays consisting of 27,648 cDNA or ESTs, and found a small heat shock protein, HSPB7, to be significantly and commonly downregulated in RCC. Subsequent quantitative PCR (qPCR) and immunohistochemical (IHC) analyses confirmed the downregulation of HSPB7 in RCC tissues and cancer cell lines in both transcriptional and protein levels. Bisulfite sequencing of a genomic region of HSPB7 detected DNA hypermethylation of some segments of HSPB7 in RCC cells and concordantly 5-aza-2'-deoxycytidine (5-Aza-dC) treatment of cancer cells restored HSPB7 expression significantly. Ectopic introduction of HSPB7 in five RCC cell lines remarkably suppressed cancer cell growth. Interestingly, we found that HSPB7 expression could be induced by p53 in a dose-dependent manner, indicating that this gene functions in the p53 pathway. Our results imply that HSBP7 is likely to be a tumor suppressor gene regulated by p53 and its downregulation by hypermethylation may play a critical role in renal carcinogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10376 CompoZr® Knockout ZFN Kit, Human HSPB7 (NM_014424)