EMAIL THIS PAGE TO A FRIEND
Reproduction (Cambridge, England)
Possible roles of the cAMP-mediators EPAC and RAP1 in decidualization of rat uterus.
PMID 24586073
Abstract
The optimal decidualization of endometrial stromal cells (ESCs) following embryo implantation is one of the critical steps to establish pregnancy in rodents and humans. This step is intricately regulated by ovarian hormones. Using in vitro human ESCs model, we previously showed that activation of a cAMP mediator, exchange protein directly activated by cAMP (EPAC), promotes ovarian steroid- or cAMP analog-induced decidualization. However, expressions and functions of EPAC and RAP1 in the uterus during pregnancy have not yet been examined. In this study, we found that the expression of EPAC2 and RAP1 was markedly upregulated in the decidual cells at the implantation sites on days 7 and 9 of pregnancy in rats. Furthermore, both delayed-implantation and artificial decidualization models showed that EPAC2 and RAP1 expression was enhanced in decidual cells. Significant activation of cAMP-responsive element-binding protein (CREB), a central transcriptional factor of cAMP signaling, was observed in decidual cells. These spatiotemporal expressions of protein related EPAC pathway are overlapped by sites with activated cAMP signaling, indicating the association of EPAC signaling with decidualization. Strikingly, further studies in in vitro rat decidualization model showed that the cAMP analog and medroxyprogesterone stimulated the expression of decidual markers, while knockdown of EPAC1/2 and RAP1 attenuated the expressions of these markers. Together, these findings suggest that EPAC and RAP1 are the crucial factors for endometrial decidualization in rat pregnancy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV40173
Anti-ACTB antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2100037
Anti-ACTB antibody produced in rabbit, affinity isolated antibody
GW23014 Anti-Actin, Cytoplasmic 1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500350
Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
R5405
Anti-Rabex-5 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3500995
Anti-RABEX5 antibody produced in rabbit, affinity isolated antibody
SAB1407359
Anti-RABGEF1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2700823
Anti-RAP1A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA006719
Anti-TERF2IP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106088
Anti-TERF2IP antibody produced in rabbit, affinity isolated antibody
SAB2107262
Anti-TERF2IP antibody produced in rabbit, affinity isolated antibody
M1629
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
C24H34O4
46412
Medroxyprogesterone 17-acetate, VETRANAL, analytical standard
C24H34O4
1378001
Medroxyprogesterone acetate, United States Pharmacopeia (USP) Reference Standard
C24H34O4
M0250000
Medroxyprogesterone acetate, European Pharmacopoeia (EP) Reference Standard
C24H34O4
M0250010
Medroxyprogesterone acetate, for performance test, European Pharmacopoeia (EP) Reference Standard
C24H34O4
Y0001484
Medroxyprogesterone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
C24H34O4
Y0000598
Medroxyprogesterone acetate for system suitability, European Pharmacopoeia (EP) Reference Standard
C24H34O4
EHU092751 MISSION® esiRNA, esiRNA human RAP1A (esiRNA1)
EHU041401 MISSION® esiRNA, esiRNA human TERF2IP (esiRNA1)
EHU120451 MISSION® esiRNA, esiRNA human RAPGEF3 (esiRNA1)
EHU080731 MISSION® esiRNA, esiRNA human RAPGEF4 (esiRNA1)
EMU053261 MISSION® esiRNA, esiRNA targeting mouse Rapgef3 (esiRNA1)
EMU047541 MISSION® esiRNA, esiRNA targeting mouse Rapgef4 (esiRNA1)
EMU092631 MISSION® esiRNA, esiRNA targeting mouse Rap1a (esiRNA1)
WH0000060M1
Monoclonal Anti-ACTB antibody produced in mouse, clone 3G4-F9, purified immunoglobulin, buffered aqueous solution
SAB1403520
Monoclonal Anti-ACTB antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution
R8154 Monoclonal Anti-RAP1 antibody produced in mouse, ~2 mg/mL, clone 4C8/1, purified immunoglobulin, buffered aqueous solution
WH0005906M1
Monoclonal Anti-RAP1A antibody produced in mouse, clone 3E1, purified immunoglobulin, buffered aqueous solution
SAB5300214
Monoclonal Anti-RAP1A antibody produced in mouse, clone 5F8, purified immunoglobulin, buffered aqueous solution