EMAIL THIS PAGE TO A FRIEND
Alzheimer's & dementia : the journal of the Alzheimer's Association
Aβ and cognitive change: examining the preclinical and prodromal stages of Alzheimer's disease.
PMID 24589436
Abstract
High β-amyloid (Aβ) is associated with faster memory decline in healthy individuals and adults with mild cognitive impairment (MCI). However, longer prospective studies are required to determine if Aβ-related memory decline continues and whether it is associated with increased rate of disease progression. Healthy controls (HCs; nxa0=xa0177) and adults with MCI (nxa0=xa048) underwent neuroimaging for Aβ and cognitive assessment at baseline. Cognition was reassessed 18 and 36 months later. Compared with low-Aβ HCs, high-Aβ HC and MCI groups showed moderate decline in episodic and working memory over 36 months. Those with MCI with low Aβ did not show any cognitive decline. Rates of disease progression were increased in the high-Aβ HC and MCI groups. In healthy individuals, high Aβ likely indicates that Alzheimer's disease (AD)-related neurodegeneration has begun. Once commenced, the rate of decline in cognitive function remains constant across the preclinical and prodromal stages of AD.
Related Materials